EasySep™ Human Eosinophil Isolation Kits
Product Details & Documents
Immunomagnetic isolation of untouched human eosinophils.
- Fast, easy-to-use and column-free
- Up to 99% purity
- Untouched, viable cells
The EasySep™ Human Eosinophil Isolation Kit is designed to isolate eosinophils from a polymorphonuclear cell-rich fraction of peripheral blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-eosinophils and magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
The kit is compatible with cells prepared using either Lymphoprep™ centrifugation (preferred for higher cell purities) or HetaSep™ sedimentation (preferred for faster, lysis-free sample processing). This product replaces the EasySep™ Human Eosinophil Enrichment Kit for even faster cell isolations.
Advarsel: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifikationer
- Beskrivelse:EasySep™ human eosinophil isolation kit, 1×10⁹ cells