Orders LinkContactUsLinkComponent
Spray Dusters, Q-Connect
Spray Dusters, Q-Connect
HFC-free spray duster, invertible, flammable, 200 ml
Katalog # DIALKF17441
Leverandør:   DIALSTAT OFFICE SUPPLIES
Spray Dusters, Q-Connect
Katalog # DIALKF17441
Spray Dusters, Q-Connect
HFC-free spray duster, invertible, flammable, 200 ml
Price
65,40 kr
Log ind for tilgængelighed
1 * 200 mL
Product Details & Documents
Air dusters for cleaning computers and electronic maintenance.
  • HFC free for environmental friendliness
  • Provides an easy way to reach out of the way areas
Specifikationer
  • Beskrivelse:
    HFC-free spray duster, invertible, flammable, 200 ml