Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Aluminium seals, lined, WHEATON®

Aluminium seals, lined, WHEATON®

Natural, center disc tear-out, PTFE/red rubber, 13 mm, aluminium
Katalog # WHEA224222-01
Leverandør:   DWK Life Sciences
Aluminium seals, lined, WHEATON®
Katalog # WHEA224222-01
Leverandør:  DWK Life Sciences
Aluminium seals, lined, WHEATON®
Aluminium seals, lined, WHEATON®

Natural, center disc tear-out, PTFE/red rubber, 13 mm, aluminium

Price
8.130,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents

Open top seal provides for filling or retrieving contents with a syringe, large target diameter.

  • For use with 12×32 mm crimp top vials
  • Choice of liner

They are also ideal for use as closures in chromatography and instrumentation applications.

Specifikationer
  • Ø:
    13 mm
  • Septum:
    PTFE/red rubber
  • Model:
    Natural, center disc tear-out
  • Materiale:
    aluminium
  • Seq:
    06
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.