Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518
gul
Katalog # SIMPM518-5T
Leverandør:   Simport Scientific
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518
Katalog # SIMPM518-5T
Leverandør:  Simport Scientific
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518
gul
Price
4.770,00 kr
Log ind for tilgængelighed
1 * 2.000 Items
Product Details & Documents
Secure specimens during processing with Swingsette™ Biopsy processing/embedding cassette, in Quickload™ stacks.
  • Anterior writing area, 45°
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
  • Large tab for convenient and easy opening
They keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximise fluid exchange and ensure proper drainage.
These cassettes are especially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets which has 1 mm square openings to maximize fluid exchange.
 : Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Specifikationer
  • Farve:
    gul
  • Seq:
    11