Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518

Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518

grøn
Catalog # SIMPM518-4T
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518
Catalog # SIMPM518-4T
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes in Quickload™ stacks, Swingsette™, M518

grøn

Price
4.770,00 kr
Log in for availability
1 * 2.000 Items
Product Details & Documents

Secure specimens during processing with Swingsette™ Biopsy processing/embedding cassette, in Quickload™ stacks.

  • Anterior writing area, 45°
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
  • Large tab for convenient and easy opening

They keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximise fluid exchange and ensure proper drainage.

These cassettes are especially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets which has 1 mm square openings to maximize fluid exchange.

Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.

Specifications
  • Farve:
    grøn
  • Seq:
    02