Quantitative filter papers, pyramid folded, Whatman™
40, 125 mm
WHAT9892-128
Easily fits into conical funnels.
- High quality and consistency
- Easy to use
- Kvalitet:40
- Seq:06
- Størrelse:125 mm
- Tykkelse:215 µm
- Cat. No.:WHAT9892-128
- Vægt:85 g/m²
- Askeindhold:0,007%