Electronic crimping tools
Electronic crimper for 13 mm Flip Top / Flip Off caps
Katalog # MANA735533
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
These battery-powered electronic crimping tools are suitable for aluminium crimp and Flip Top / Flip Off caps (not suitable for 20 mm magnetic / bi-metal crimp caps).
- Mobile tools for consistent and reproducible crimping results
- Crimping pressure adjustable by the control unit on top of the tool
- Long lasting lithium ion cell batteries (full battery charge for several hundred vials, life time of battery >1500 charges)
Certifications: CE certificate of conformity along with one year warranty.
Documents
Brochure-Crimping-tools-EN.pdf Brochure-Vials-and-caps-EN.pdf
Yderligere produktdetaljer
Specifikationer
- Beskrivelse:Electronic crimper for 13 mm Flip Top / Flip Off caps
- Seq:03
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Rack for 2 electronic crimping tools
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement battery 6,6 V, 8,6 Wh