Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Assistent® Brushes with Cotton Tip Bristles

Assistent® Brushes with Cotton Tip Bristles

Bomuld, 8 mm, Wire, 150 mm
Katalog # HECH91120000001
Assistent® Brushes with Cotton Tip Bristles
Katalog # HECH91120000001
Assistent® Brushes with Cotton Tip Bristles
Assistent® Brushes with Cotton Tip Bristles

Bomuld, 8 mm, Wire, 150 mm

Price
1.390,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents

Brushes, light bristles, with cotton tip.

Specifikationer
  • Børstemateriale:
    Bomuld
  • Børstehoved Ø:
    8 mm
  • Handle material:
    Wire
  • Seq:
    02
  • Samlet længde:
    150 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.