Labels for BradyJet J2000 Lab printer
Printer labels, B-7425J, PP, hvid, J2000-Lab, 25×9 mm, 2000 labels/roll
Katalog # BRDY150005
Leverandør: Brady
Gå til:
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Specifikationer
- Printer:J2000-Lab
- Model:B-7425J
- Farve:hvid
- Beskrivelse:Printer labels
- Dimensioner:25×9 mm
- Materiale:PP
- Seq:01
- Pakket:2000 labels/roll