Volumetric flasks, class A, white markings
2 ml, 10/19, 0,04 ± ml
Katalog # WITG3.669.001.10BR
Leverandør: witeg Labortechnik
Volumetric flasks, class A, white markings
Katalog # WITG3.669.001.10BR
Leverandør: witeg Labortechnik
Gå til:
Product Details & Documents
Borosilicate glass 3.3, amber, with glass stopper.
- White markings
- Conformity certified (DE-M marking)
- Inclusive batch certificate
Certifications: DIN EN ISO 1042
Specifikationer
- NS:10/19
- Kapacitet:2 ml
- Tolerance:0,04 ± ml