Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
2 ml, 10/19, 0,04 ± ml
Katalog # WITG3.669.001.10BR
Leverandør:   witeg Labortechnik
Volumetric flasks, class A, white markings
Katalog # WITG3.669.001.10BR
Leverandør:  witeg Labortechnik
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
2 ml, 10/19, 0,04 ± ml
Price
351,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3, amber, with glass stopper.
  • White markings
  • Conformity certified (DE-M marking)
  • Inclusive batch certificate
Certifications: DIN EN ISO 1042
Specifikationer
  • NS:
    10/19
  • Kapacitet:
    2 ml
  • Tolerance:
    0,04 ± ml