WATER DENSITY/REFRACTIVE INDEX COM STD 1 * 1 Items
Katalog # METT51338010
Leverandør: Mettler - Toledo
WATER DENSITY/REFRACTIVE INDEX COM STD 1 * 1 Items
Katalog # METT51338010
Leverandør: Mettler - Toledo
Frequently Bought Together
VWR®, Standards for water determination by the Karl Fischer method
Leverandør: VWR Chemicals
VWR®, Men's Coats, Cotton or Polyester/Cotton
Leverandør: VWR Collection
VWR®, Manual Crimper and Decapper
Leverandør: VWR Collection
VWR® pH Buffer Solution According to Reag. Ph. Eur. 4000600
Leverandør: VWR Chemicals
Kondensator med kølespiral og kappe, med standard jordforbindelser
Leverandør: Lenz Laborglas GmbH & CO.KG
Glycerin 85% Ph. Eur.
Leverandør: VWR Chemicals
Natriumcarbonat, vandfri, Suprapur® til sporstofanalyse, Supelco®
Leverandør: Merck
VWR®, Disposable Micro Weighing Dishes
Leverandør: VWR Collection
Hexamethylenetetramin ≥99.0%, AnalaR NORMAPUR® analytical reagent
Leverandør: VWR Chemicals
Repetitive pipette, HandyStep® touch and HandyStep® touch S
Leverandør: Brand
Bottles, wide neck, with screw cap
Leverandør: LAMAPLAST
Combined density and refractometry standards
Leverandør: Mettler - Toledo
VWR®, Density Standards, Certified Reference Materials, Referenced at 20 °C
Leverandør: VWR Chemicals
pH-elektroder, InLab®, Expert-serien
Leverandør: Mettler - Toledo
Trimethylolpropane triacrylate stabilised
Leverandør: Thermo Fisher Scientific
[EN]ACRYLAMIDE-2,3,3 D3 1 * 10 mg
Leverandør: EHRENSTORFER
Silica ≥99.8%, Supelco®
Leverandør: Merck