Orders LinkContactUsLinkComponent
Accessories for circulators
Accessories for circulators
4-way header with ball valves, M16×1, The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.
Katalog # HUBE343304
Leverandør:   HUBER
Accessories for circulators
Katalog # HUBE343304
Leverandør:  HUBER
Accessories for circulators
4-way header with ball valves, M16×1, The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.
Price
4.740,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    4-way header with ball valves, M16×1
  • Seq:
    05
  • Til:
    The 4-way header with universal mounting bracket for wall mounting the manifold. A suitable length hose is required to connect the manifold and the temperature control unit. Note: The manifold must not be mounted on the unit.