Orders LinkContactUsLinkComponent
VWR® Evaporating Flasks
VWR® Evaporating Flasks
1000 ml, 29/32 NS
Katalog # 531-1102
Leverandør:   VWR Collection
VWR® Evaporating Flasks
Katalog # 531-1102
Leverandør:  VWR Collection
VWR® Evaporating Flasks
VWR® Evaporating Flasks
1000 ml, 29/32 NS
Price
428,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance and high resistance to thermal shock
  • White labelling area and imprinted catalogue number
These pear-shaped rotary evaporating flasks are produced from thick walled borosilicate glass tubing. With standard ground socket, suitable for all standard rotary evaporators.
Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    1000 ml
  • Seq:
    04