Orders LinkContactUsLinkComponent
VWR®, UV/Visible standards
VWR®, UV/Visible standards
UV/Vis standard 3, for stray light testing, 1 KIT
Katalog # 86127.001
Leverandør:   VWR Chemicals
VWR®, UV/Visible standards
Katalog # 86127.001
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, UV/Visible standards
VWR®, UV/Visible standards
UV/Vis standard 3, for stray light testing, 1 KIT
CAS nummer:  
Price
1.740,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
For spectrophotometer calibration according to European Pharmacopoeia (Ph. Eur.)
  • Absorbance control
  • Testing of resolution power
  • Stray light testing
  • Wavelength accuracy testing
In the pharmaceutical industry, a regular check of UV/Visible spectrophotometers according to Ph. Eur. is mandatory. Even in other industries, researchers who are using UV/Visible spectrophotometers, and who are working according to e.g. GMP, GLP, USP or ISO guidelines, need to check and qualify their instruments on a regular basis.
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    UV/Vis standard 3, for stray light testing
Specifikationer