Orders LinkContactUsLinkComponent
Accessories for CryoPlus Sample Storage Systems
Rack for Matrix 2D plates measuring 58,42 mm tall, 572×135×93 mm, for CryoPlus 1, 2, 4
Katalog # BARN1950834
Leverandør:   Thermo Fisher Scientific
Accessories for CryoPlus Sample Storage Systems
Katalog # BARN1950834
Accessories for CryoPlus Sample Storage Systems
Rack for Matrix 2D plates measuring 58,42 mm tall, 572×135×93 mm, for CryoPlus 1, 2, 4
Price
kr1,320.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Rack for Matrix 2D plates measuring 58,42 mm tall, 572×135×93 mm, for CryoPlus 1, 2, 4
  • Seq:
    12