Accessories for CryoPlus Sample Storage Systems
Rack for Matrix 2D plates measuring 58,42 mm tall, 572×135×93 mm, for CryoPlus 1, 2, 4
Katalog # BARN1950834
Leverandør: Thermo Fisher Scientific
Accessories for CryoPlus Sample Storage Systems
Katalog # BARN1950834
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Rack for Matrix 2D plates measuring 58,42 mm tall, 572×135×93 mm, for CryoPlus 1, 2, 4
- Seq:12