Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld

VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld

Digital handheld phenomenal® MD 8000 H multi-parameter meter, dissolved oxygen kit, with intelligent optical D.O./temperature sensor OPOX 11-3
Katalog # 665-0496
Leverandør:   VWR Collection
VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld
Katalog # 665-0496
Leverandør:  VWR Collection
VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld
VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld

Digital handheld phenomenal® MD 8000 H multi-parameter meter, dissolved oxygen kit, with intelligent optical D.O./temperature sensor OPOX 11-3

Price
15.070,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter and no need to adjust the meter’s settings). The measured data is complete with serial number, settings and calibration data.

  • One universal measuring channel
  • Digital sensor recognition: The measured parameter and settings are recognised automatically
  • Suitable for Intelligent Digital Probes (IDP)
  • Suitable for measurements with cable length up to 100 metres
  • 4500-point memory
  • Easy to read backlit B/W graphic display
  • Low energy consumption (1000 operating hours)

Ordering information: Additional probes for other parameters can be added. Please contact your local Avantor sales office for further information on other electrodes available.

Delivery information: 665-0494 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal IDP 7211 and USB cable.
665-0495 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml conductivity standard 0,01 mol/L KCl, conductivity cell IDP 761-C and USB cable.
665-0496 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, optical DO sensor OPOX11-3 and USB cable.

Specifikationer
  • Beskrivelse:
    Digital handheld phenomenal® MD 8000 H multi-parameter meter, dissolved oxygen kit, with intelligent optical D.O./temperature sensor OPOX 11-3
  • mV - Område:
    ±2500; ±1200
  • Konduktivitetscellekonstant:
    Automatic
  • Konduktivitetsområde:
    0,01 µS/cm - 1000 mS/cm (depending on sensor)
  • Referencetemperatur:
    20/25 °C
  • IP rating:
    IP 67
  • Kalibrering:
    Anycal 5-point customer buffer calibration
  • Datalog Kapacitet:
    Up to 4500 data sets in automatic
  • Tilslutninger:
    1 measurement input
  • Kalibreringspunkter:
    1 to 5 (automatic buffer recognition)
  • D.O. - mætning:
    0,0 - 200,0; 0 - 500 %
  • mV - Aflæsningsinterval:
    0,1; 1
  • Konduktivitetsnøjagtighed:
    ±0,5% of value
  • mV - Nøjagtighed:
    ±0,3; ±1
  • pH - Aflæsningsinterval:
    0,1/0,01/0,001
  • Salinitet - Område:
    0 - 70
  • Hukommelse:
    4500 points
  • Datalog - Log-funktion:
    Manual/time controlled
  • pH - Nøjagtighed:
    ±0,1, ±0,01, ±0,005
  • Temperaturinterval:
    Auto: −5...+105 °C
  • TDS - Område:
    0 mg/L - 199,9 g/L
  • pH - Område:
    0 - 14
  • DO Koncentration:
    0,00 - 20,00 / 0,0 - 50,0 mg/L
  • Temperature resolution:
    0,1 °C
  • Temperature accuracy:
    ±0,2 K
  • Modstand - Område:
    1 Ωcm - 20 MΩcm (depending on sensor)
  • Cat. No.:
    665-0496
  • Parameter:
    pH, mV, temperature, conductivity, dissolved oxygen, turbidity
  • Interface:
    Mini USB-B (USB 1.1) for data downloads
  • Display:
    Blacklit B/W graphic LCD
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.