VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld
Product Details & Documents
These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter and no need to adjust the meter’s settings). The measured data is complete with serial number, settings and calibration data.
- One universal measuring channel
- Digital sensor recognition: The measured parameter and settings are recognised automatically
- Suitable for Intelligent Digital Probes (IDP)
- Suitable for measurements with cable length up to 100 metres
- 4500-point memory
- Easy to read backlit B/W graphic display
- Low energy consumption (1000 operating hours)
Bestillingsoplysninger: Additional probes for other parameters can be added. Please contact your local Avantor sales office for further information on other electrodes available.
Leveringsinformation: 665-0494 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal IDP 7211 and USB cable.
665-0495 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml conductivity standard 0,01 mol/L KCl, conductivity cell IDP 761-C and USB cable.
665-0496 includes meter, case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, optical DO sensor OPOX11-3 and USB cable.
Specifikationer
- Beskrivelse:Digital handheld phenomenal® MD 8000 H multi-parameter meter, conductivity kit, with intelligent conductivity/temperature cell IDP 761-C with 1 bottle of 50 ml conductivity standard (1413 µS)
- mV - Område:±2500; ±1200
- Konduktivitetscellekonstant:Automatic
- Konduktivitetsområde:0,01 µS/cm - 1000 mS/cm (depending on sensor)
- Referencetemperatur:20/25 °C
- IP rating:IP 67
- Kalibrering:Anycal 5-point customer buffer calibration
- Datalog Kapacitet:Up to 4500 data sets in automatic
- Tilslutninger:1 measurement input
- Kalibreringspunkter:1 to 5 (automatic buffer recognition)
- D.O. - mætning:0,0 - 200,0; 0 - 500 %
- mV - Aflæsningsinterval:0,1; 1
- Konduktivitetsnøjagtighed:±0,5% of value
- mV - Nøjagtighed:±0,3; ±1
- pH - Aflæsningsinterval:0,1/0,01/0,001
- Salinitet - Område:0 - 70
- Hukommelse:4500 points
- Datalog - Log-funktion:Manual/time controlled
- pH - Nøjagtighed:±0,1, ±0,01, ±0,005
- Temperaturinterval:Auto: −5...+105 °C
- TDS - Område:0 mg/L - 199,9 g/L
- pH - Område:0 - 14
- DO Koncentration:0,00 - 20,00 / 0,0 - 50,0 mg/L
- Temperature resolution:0,1 °C
- Temperature accuracy:±0,2 K
- Modstand - Område:1 Ωcm - 20 MΩcm (depending on sensor)
- Cat. No.:665-0495
- Parameter:pH, mV, temperature, conductivity, dissolved oxygen, turbidity
- Interface:Mini USB-B (USB 1.1) for data downloads
- Display:Blacklit B/W graphic LCD



