Wringers for mops
Stainless steel curved mop wringer for EasyCurve™ mops, for 20 l buckets, 546×146×64 mm
Catalog # CONT2694
Supplier: CONTEC
Jump to:
Product Details & Documents
Variety of wringers for curved and flat mops.
- Made of stainless steel
- Can be used with curved, flat, and string mops
- Ideal for use with Contec’s 15, 20, and 25 L bucket systems
- Fully autoclavable
Specifications
- Beskrivelse:Stainless steel curved mop wringer for EasyCurve™ mops, for 20 l buckets
- Dimensioner:546×146×64 mm
- Seq:02