Wipes, Amplitude™ Sigma™ Plus
Amplitude™ Sigma™ Plus wipes, flat stacked, hvid, 305×305 mm
Katalog # CONTC1E-1212
Leverandør: CONTEC
Gå til:
Product Details & Documents
The Amplitude™ Sigma™ Plus wipes are strong and durable, compatible with most solvents, and have outstanding sorbency.
- Hydroentangled cellulose/poylester provides excellent strength and durability
- Low levels of particles and extractables
- Variety of blended fibers enable excellent absorbing capacity
Amplitude™ Sigma™ Plus Wipes are constructed using a nonwoven fabric that is a hydroentangled blend of polyester and cellulose fibers. Wet or dry, the wipe maintains a high level of strength and durability.
Specially treated to reduce particles and fibers, it is recommended when low particle generation combined with sorbency is of high priority.
Specifikationer
- Farve:hvid
- Beskrivelse:Amplitude™ Sigma™ Plus wipes, flat stacked
- L×B:305×305 mm
- Seq:02