Orders LinkContactUsLinkComponent
Re-Usable Cleaning Coupons
Borosilicate glass, 100×100 mm
  SAMP8644AB-100
Re-Usable Cleaning Coupons
  SAMP8644AB-100
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons
Borosilicate glass, 100×100 mm
887,00 kr
1 * 1 Items
Cleaning coupons are great for validating a cleaning regime.
  • All popular sizes available
  • Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
  • Beskrivelse:
    Borosilicate glass
  • L×B:
    100×100 mm
  • Seq:
    06