Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Empty FPLC columns, G-TRAP™

Empty FPLC columns, G-TRAP™

PP, 1 ml
Katalog # GENO786-1290
Leverandør:   G-Biosciences
Empty FPLC columns, G-TRAP™
Katalog # GENO786-1290
Leverandør:  G-Biosciences
Empty FPLC columns, G-TRAP™
Empty FPLC columns, G-TRAP™

PP, 1 ml

Price
232,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These empty columns are designed for fast protein liquid chromatography.

  • Frit material is ultra-high molecular weight polyethylene

Columns are made of polypropylene and come in 1 ml and 5 ml sizes.

 : Includes column tube, sealing plug, sealing sleeve, top stop plug and bottom stop plug, top (large) frit, bottom (small) frit (inserted in column).

Specifikationer
  • Kapacitet:
    1 ml
  • Seq:
    01
  • Kolonnemateriale:
    PP
  • Cat. No.:
    GENO786-1290
  • Maks. tryk:
    3 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.