Optimum Growth Flask
Optimum Growth™ flask, 2,8 L
Leverandør: THOMSON INSTRUMENT
Gå til:
Product Details & Documents
Optimum Growth™ flasks (patented) give excellent growth and expression for both mammalian and insect cell lines.
- Baffles designed for high aeration and low shear
- Same footprint as compairable Fernbach flask
- Less foaming than disposable Fernbach flask
- Transfer cap connects directly to Wave Bags™ and bioreactors with quick connect or tube fusing
- 0,2 μm vented cap
- Individually packaged and sterilised
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Kapacitet:2,8 L
- Beskrivelse:Optimum Growth™ flask
- Seq:6
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



