Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Depth filter capsules, Supracap™ 100

Depth filter capsules, Supracap™ 100

Seitz® PDP8 media with 1 - 1-1/2" hose barb inlet/outlet, 6,0 - 30,0 µm, 1000 cm²
Katalog # PALLNP6PDP81
Depth filter capsules, Supracap™ 100
Katalog # PALLNP6PDP81
Depth filter capsules, Supracap™ 100
Depth filter capsules, Supracap™ 100

Seitz® PDP8 media with 1 - 1-1/2" hose barb inlet/outlet, 6,0 - 30,0 µm, 1000 cm²

Price
3.500,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Supracap™ 100 depth filter capsules with PP housing are scalable capsules with Seitz® high performance depth filter media. They are suitable for process development and pilot scale applications.

  • Seamless linear scalability
  • Completely disposable
  • Encapsulated design reduces operator exposure to potential biohazards

Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.

Applications include biopharmaceuticals, mammalian cell cultures, therapeutic protein, vaccines, blood plasma proteins and serum, yeast, E.coli lysates and refolds.

The Seitz® Bio 20 media grade is made of highly purified natural and modified celluloses and is free from inorganic materials. Seitz® Bio 20 media is ideal for ion sensitive products such as therapeutic proteins, dialysis solutions, and protein-based diagnostics.

Seitz® P-series media are comprised of cellulose fibers, filter aids (diatomaceous earth and perlite), and resins. This grade is recommended for coarse filtration in the biotechnological and pharmaceutical industries.

Seitz® HP-series media are feature two distinct layers of Seitz® P-series depth filter sheets that feature a more permeable layer followed by a less permeable layer (PDD1, PDE2, PDH4 and PDK5). Seitz® HP-series media performs exceptionally well in low viability, high solids-containing applications, and with wider distribution of particle sizes in biotech applications.

Certifications: Manufactured under a quality management system certified to ISO 9000 and ISO 9001, and an environmental management system certified to ISO 14001.

Specifikationer
  • Filtreringsområde:
    1000 cm²
  • Porestørrelse:
    6,0 - 30,0 µm
  • Model:
    Seitz® PDP8 media with 1 - 1-1/2" hose barb inlet/outlet
  • Seq:
    38
  • Cat. No.:
    PALLNP6PDP81
  • Max. temperature:
    40 °C
  • Maks. tryk:
    3 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.