Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Depth filter capsules, Supracap™ 100

Depth filter capsules, Supracap™ 100

Seitz® PDE2 media with 1 - 1-1/2" sanitary flange connections, 154×213 mm, 0,2 - 3,5 µm, 250 cm²
Katalog # PALLNP5LPDE21
Depth filter capsules, Supracap™ 100
Katalog # PALLNP5LPDE21
Depth filter capsules, Supracap™ 100
Depth filter capsules, Supracap™ 100

Seitz® PDE2 media with 1 - 1-1/2" sanitary flange connections, 154×213 mm, 0,2 - 3,5 µm, 250 cm²

Price
2.020,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Supracap™ 100 depth filter capsules with PP housing are scalable capsules with Seitz® high performance depth filter media. They are suitable for process development and pilot scale applications.

  • Seamless linear scalability
  • Completely disposable
  • Encapsulated design reduces operator exposure to potential biohazards

Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.

Applications include biopharmaceuticals, mammalian cell cultures, therapeutic protein, vaccines, blood plasma proteins and serum, yeast, E.coli lysates and refolds.

The Seitz® Bio 20 media grade is made of highly purified natural and modified celluloses and is free from inorganic materials. Seitz® Bio 20 media is ideal for ion sensitive products such as therapeutic proteins, dialysis solutions, and protein-based diagnostics.

Seitz® P-series media are comprised of cellulose fibers, filter aids (diatomaceous earth and perlite), and resins. This grade is recommended for coarse filtration in the biotechnological and pharmaceutical industries.

Seitz® HP-series media are feature two distinct layers of Seitz® P-series depth filter sheets that feature a more permeable layer followed by a less permeable layer (PDD1, PDE2, PDH4 and PDK5). Seitz® HP-series media performs exceptionally well in low viability, high solids-containing applications, and with wider distribution of particle sizes in biotech applications.

 : Manufactured under a quality management system certified to ISO 9000 and ISO 9001, and an environmental management system certified to ISO 14001.

Specifikationer
  • Filtreringsområde:
    250 cm²
  • Porestørrelse:
    0,2 - 3,5 µm
  • Model:
    Seitz® PDE2 media with 1 - 1-1/2" sanitary flange connections, 154×213 mm
  • Seq:
    06
  • Cat. No.:
    PALLNP5LPDE21
  • Max. temperature:
    40 °C
  • Maks. tryk:
    3 bar