Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(R)-(-)-Hydratropasyre 98%

(R)-(-)-Hydratropasyre 98%

Bottle, 5 g
Katalog # APOSOR304085-5G
Leverandør:   Apollo Scientific
(R)-(-)-Hydratropasyre 98%
Katalog # APOSOR304085-5G
Leverandør:  Apollo Scientific
CAS nummer:  7782-26-5
(R)-(-)-Hydratropasyre 98%

Bottle, 5 g

CAS nummer:  7782-26-5
Price
242,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
  • CAS nummer:7782-26-5
  • Formel:C₉H₁₀O₂
  • MDL nummer:MFCD00063140
  • Molecular Weight:150.18 g/mol
  • Smeltende Pt:29…30 °C
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.