(R)-(-)-Hydratropasyre 98%
Bottle, 25 g
Katalog # APOSOR304085-25G
Leverandør: Apollo Scientific
(R)-(-)-Hydratropasyre 98%
Katalog # APOSOR304085-25G
Leverandør: Apollo Scientific
CAS nummer: 7782-26-5
Gå til:
Product Details & Documents
Synonymer: (R)-(-)-2-Phenylpropanoic acid, (R)-(-)-2-Phenylpropionic acid, (R)-2-Phenylpropanoic acid
- CAS nummer:7782-26-5
- Formel:C₉H₁₀O₂
- MDL nummer:MFCD00063140
- Molecular Weight:150.18 g/mol
- Smeltende Pt:29…30 °C
- Opbevaringstemperatur:Stue temperatur
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:25 g



