Orders LinkContactUsLinkComponent
Hæmatoxylin
Bottle, 50 g
  APOSBIH6002-50G
Hæmatoxylin
  APOSBIH6002-50G
 :  517-28-2
Hæmatoxylin
Bottle, 50 g
 :  517-28-2
1.820,00 kr
1 * 50 g
 :  DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin
  •  :517-28-2
  •  :C₁₆H₁₄O₆
  •  :MFCD00078111
  •  :302.28 g/mol
  •  :200 °C (dec.)
  •  :Stue temperatur
  •  :208-237-3
  • Pakning:
    Bottle
  • Pakn:
    50 g