Orders LinkContactUsLinkComponent
SeramunBlock
SeramunBlock, 250 mL
Katalog # APOSBIL3122-250ML
Leverandør:   Apollo Scientific
SeramunBlock
Katalog # APOSBIL3122-250ML
Leverandør:  Apollo Scientific
CAS nummer:  
SeramunBlock
SeramunBlock, 250 mL
CAS nummer:  
Price
747,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents
Combined Stabiliser/ blocking solution for solid phase immobilised proteins effecting protein stabilisation and blocking of free surfaces.
Specifikationer
Specifikationer
  • Pakn:
    250 mL
  • Beskrivelse:
    SeramunBlock