Kanelaldehyd 98%
Bottle, 500 g
Katalog # APOSOR55404-500G
Leverandør: Apollo Scientific
Gå til:
Product Details & Documents
Synonymer: trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
- CAS nummer:104-55-2
- Formel:C₉H₈O
- MDL nummer:MFCD00007000
- Molecular Weight:132.16 g/mol
- :Density: 1.112 g/cm³ (20 °C)
- Opbevaringstemperatur:Fryser
Specifikationer
Specifikationer
- Pakning:Bottle
- Pakn:500 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



