Objektglas med fordybning
Microscope slides with reaction wells, blå, 10, 7 mm
Catalog # MARI1216531
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Documents
Specifications
- Ant. brønde:10
- Farve:blå
- Beskrivelse:Microscope slides with reaction wells
- Seq:27
- Brønd Ø:7 mm