Orders LinkContactUsLinkComponent
Detergent, Contec® Sterile NeutraKlean
Detergent, Contec® Sterile NeutraKlean
Contec® Sterile NeutraKlean
Neutral detergent with purified water, capped, 1 litre capped container
Katalog # CONTSBC103NK
Leverandør:   CONTEC
Detergent, Contec® Sterile NeutraKlean
Katalog # CONTSBC103NK
Leverandør:  CONTEC
Detergent, Contec® Sterile NeutraKlean
Detergent, Contec® Sterile NeutraKlean
Contec® Sterile NeutraKlean
Neutral detergent with purified water, capped, 1 litre capped container
Price
2.180,00 kr
Log ind for tilgængelighed
1 * 6 Items
Product Details & Documents
Contec® Sterile NeutraKlean is a gentle and low-foaming detergent designed for the cleaning of life science cleanrooms. Ready to use, irradiated neutral detergent with purified water.
  • Neutral pH makes it suitable for use in all areas of cleanroom as gentle on surfaces
  • Filtered to 0.2 microns under Grade A airflow in an Grade C cleanroom ensures the product is free from contamination and particulates
  • Low-foaming detergent action is ideal for use on floors, walls, and benches
  • Low level of surfactant provides low residue which can be easily removed
  • Trigger spray can be set to jet or spray and provides good surface coverage
  • Trigger spray and “bag in bottle” protected system prevents bottle contents from becoming contaminated during use
  • Bottles can be completely emptied eliminating wastage so no need to discard unused product during shelf life
  • Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
With a pH after irradiation between 6.5 and 8.0, Contec Sterile NeutraKlean is ideal for carrying out the low-level cleaning required after maintenance and to remove disinfectant residues and general soil. The use of a surfactant/detergent helps to break the binding of particles and dirt to a surface and allows them to be more easily removed.
The bottles are provided double bagged to aid the transfer of the product into a pharmaceutical cleanroom, isolators, or RABS. The linear tear bags are easy to open even when wearing gloves.
Bestillingsoplysninger: The product is also available non-sterile.
Leveringsinformation: The product is available as either a 1 litre trigger spray or capped bottle and a 5 litre capped container for larger areas
Emballering: Double bagged, packed in linear tear packaging.
Specifikationer
  • Beskrivelse:
    Contec® Sterile NeutraKlean
    Neutral detergent with purified water, capped
  • Seq:
    01
  • Pakket:
    1 litre capped container