Orders LinkContactUsLinkComponent
Accessories for compact bucket systems
Irradiated bucket liners, 0,1 mm polyethylene, for 15 L buckets
Katalog # 141-1438
Leverandør:   CONTEC
Accessories for compact bucket systems
Katalog # 141-1438
Leverandør:  CONTEC
Accessories for compact bucket systems
Accessories for compact bucket systems
Irradiated bucket liners, 0,1 mm polyethylene, for 15 L buckets
Price
3.470,00 kr
Log ind for tilgængelighed
1 * 60 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Irradiated bucket liners, 0,1 mm polyethylene, for 15 L buckets
  • Seq:
    03