Objectives for REVOLVE 3M microscope
Achromat objective, 2×, 5,8 mm
Katalog # ECHLJMAG0021
Leverandør: DISCOVER ECHO
Gå til:
Product Details & Documents
Documents
ECHO Meet Revolve
Produktlitteratur
Specifikationer
- Forstørrelse:2×
- Beskrivelse:Achromat objective
- Seq:03
- Arbejdsafstand:5,8 mm