Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat olie; 1,25 NA, 100×, 0,15 mm
Katalog # ECHLJMAG1005
Leverandør:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Katalog # ECHLJMAG1005
Leverandør:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat olie; 1,25 NA, 100×, 0,15 mm
Price
4.690,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Forstørrelse:
    100×
  • Beskrivelse:
    Plan achromat olie; 1,25 NA
  • Seq:
    26
  • Arbejdsafstand:
    0,15 mm