Objectives for REVOLVE 3M microscope
Plan achromat LWD fase Ph2; 0,55 NA, 40×, 2,2 mm
Catalog # ECHLJMAG0409
Supplier: DISCOVER ECHO
Jump to:
Product Details & Documents
Documents
ECHO Meet Revolve
Product Literature
Specifications
- Forstørrelse:40×
- Beskrivelse:Plan achromat LWD fase Ph2; 0,55 NA
- Seq:20
- Arbejdsafstand:2,2 mm