Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat fase Ph2; 0,65 NA, 40×, 0,6 mm
Katalog # ECHLJMAG0408
Leverandør:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Katalog # ECHLJMAG0408
Leverandør:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Plan achromat fase Ph2; 0,65 NA, 40×, 0,6 mm
Price
4.980,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Forstørrelse:
    40×
  • Beskrivelse:
    Plan achromat fase Ph2; 0,65 NA
  • Seq:
    19
  • Arbejdsafstand:
    0,6 mm