Orders LinkContactUsLinkComponent
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Fluorite phase objective, 40×, 0,51 mm
Catalog # 630-3062
Supplier:   DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Catalog # 630-3062
Supplier:  DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Fluorite phase objective, 40×, 0,51 mm
Price
14.470,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Documents
Specifications
  • Forstørrelse:
    40×
  • Beskrivelse:
    Fluorite phase objective
  • Seq:
    11
  • Arbejdsafstand:
    0,51 mm