Orders LinkContactUsLinkComponent
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat SLWD objective, 20×, 25 mm
Katalog # ECHLJMAG0208
Leverandør:   DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Katalog # ECHLJMAG0208
Leverandør:  DISCOVER ECHO
Objectives for REVOLVE 3M microscope
Objectives for REVOLVE 3M microscope
Achromat SLWD objective, 20×, 25 mm
Price
23.930,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Forstørrelse:
    20×
  • Beskrivelse:
    Achromat SLWD objective
  • Seq:
    14
  • Arbejdsafstand:
    25 mm