[EN]PIPET 5ML SER. PLAS/PLAS TRIPLE PACK 1 * 100 Items
Katalog # GREI606160-TRI_100
Leverandør: Greiner Bio-One
[EN]PIPET 5ML SER. PLAS/PLAS TRIPLE PACK 1 * 100 Items
Katalog # GREI606160-TRI_100
Leverandør: Greiner Bio-One
Frequently Bought Together
Medieflasker, firkantede, sterile, Nalgene®
Leverandør: Thermo Fisher Scientific
VWR® Ultra-High Performance Centrifuge Tubes
Leverandør: VWR Collection
VWR®, Multibrøndsplader til celledyrkning
Leverandør: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Leverandør: Corning
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Leverandør: EPPENDORF
Eppendorf® Protein LoBind® mikrocentrifugerør
Leverandør: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Leverandør: VWR Collection
VWR®, Sampling Spoons
Leverandør: VWR Collection
VWR® Premium Line, Serological Pipettes
Leverandør: VWR Collection
VWR® Blender Bags
Leverandør: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Leverandør: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Leverandør: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Leverandør: VWR Collection
VWR®, Transfer Pipettes, High Performance
Leverandør: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Leverandør: VWR Collection
VWR®, Transfer Pipettes, Graduated
Leverandør: VWR Collection