Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Adapters for universal centrifuges

Adapters for 15 ml rotor bores or adapters
  SIMPT330AD15
Adapters for universal centrifuges
  SIMPT330AD15
Adapters for universal centrifuges
Adapters for universal centrifuges

Adapters for 15 ml rotor bores or adapters

 
 
   
1 * 8 Items
 

These universal centrifuge adapters are made of acetal plastic.

  • Reusable and freezable (–90 °C)
 
  • Beskrivelse:
    Adapters for 15 ml rotor bores or adapters
  • Seq:
    01
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.