CapLock™ clips
CapLock™ clip, hvid
Katalog # SIMPT330-75CLW
Leverandør: Simport Scientific
Gå til:
Product Details & Documents
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
- Ideal for shipping samples
- Suitable for 5 ml tubes with snap cap
Specifikationer
- Farve:hvid
- Beskrivelse:CapLock™ clip
- Seq:68