Absorbent socs, Allwik Universal
43 L, 12 pieces/box, 76×122 mm
Catalog # BRDY813789
Supplier: Brady
Jump to:
Product Details & Documents
These Allwik-Socs are used for absorbing leaks and spills around machines.
- Absorbs a variety of industrial liquids including aggressive chemicals
- Flexible design allows the soc to fit around any machine, corner or curve
- Filled with universal meltblown polypropylene
Specifications
- Kapacitet:43 L
- Seq:01
- Pakket:12 pieces/box
- Ø×L:76×122 mm