Orders LinkContactUsLinkComponent
Cold Resistant Gloves, Cryo Gloves
Mid-arm length for handling samples from ultra low temperature and cryogenic storage, M, blå, 356 mm
  HERA189561
Cold Resistant Gloves, Cryo Gloves
  HERA189561
Cold Resistant Gloves, Cryo Gloves
Cold Resistant Gloves, Cryo Gloves
Mid-arm length for handling samples from ultra low temperature and cryogenic storage, M, blå, 356 mm
2.390,00 kr
1 * 1 Items
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
  • Comfortable, lightweight, breathable protection
  • Side release buckles for quick on/off
 : Available in a variety of sizes.
  • Handske størrelse:
    M
  • Farve:
    blå
  • Beskrivelse:
    Mid-arm length for handling samples from ultra low temperature and cryogenic storage
  • Længde:
    356 mm
  • Seq:
    01
  • Cat. No.:
    HERA189561
  • Ambidextrous/Hånd-specifik:
    Hånd-specifik