Cold Resistant Gloves, Cryo Gloves
Mid-arm length for handling samples from ultra low temperature and cryogenic storage, M, blå, 356 mm
HERA189561
Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.
- Comfortable, lightweight, breathable protection
- Side release buckles for quick on/off
: Available in a variety of sizes.
- Handske størrelse:M
- Farve:blå
- Beskrivelse:Mid-arm length for handling samples from ultra low temperature and cryogenic storage
- Længde:356 mm
- Seq:01
- Cat. No.:HERA189561
- Ambidextrous/Hånd-specifik:Hånd-specifik