VWR®, Levelling Platform with Bubble Level
Levelling platform with 4 independent screws and bubble level, PerfectBlue™ Mini and Midi Gel systems
Katalog # 700-0846
Leverandør: VWR Collection
Gå til:
Product Details & Documents
The use of levelling platforms in electrophoresis can be a big help in maintaining a uniform gel thickness and therefore a uniform and quality sample separation.
- Uniform gel thickness
- Consistent buffer level over the gel
- Assures high quality separation results
The levelling platform offers 4 independent screws and an easy to read bubble level to produce a level and stable platform. With a usable surface of 36×46 cm, this platform is suitable for use with both the Mini and Midi Gel systems of the PerfectBlue™ range.
Specifikationer
- Til:PerfectBlue™ Mini and Midi Gel systems
- Beskrivelse:Levelling platform with 4 independent screws and bubble level
- Seq:02