Orders LinkContactUsLinkComponent
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons
Polypropylene, 100×100 mm
Katalog # SAMP8644PP-100
Leverandør:   Sampling Systems
Re-Usable Cleaning Coupons
Katalog # SAMP8644PP-100
Leverandør:  Sampling Systems
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons
Polypropylene, 100×100 mm
Price
417,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cleaning coupons are great for validating a cleaning regime.
  • All popular sizes available
  • Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
Specifikationer
  • Beskrivelse:
    Polypropylene
  • L×B:
    100×100 mm
  • Seq:
    11