Natriumazid 99+%
Plastic bottle, 1 kg
J21610.A1
: Sodium triazide
- :26628-22-8
- :NaN₃
- :MFCD00003536
- :65.01 g/mol
- :14,08581
- :∼275 °C (dec.)
- :1,846 g/cm³
- :Stue temperatur
- :247-852-1
- :1687
- :6.1,II
Documents
- Form:White crystalline powder
- Assay (Unspecified):≥99.0%
- Loss on Drying:≤0.5%
- Pakning:Plastic bottle
- Pakn:1 kg
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



