Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Detergents

VISOCOLOR® Colorimetric Test, Detergents

VISOCOLOR® ECO Cationic detergents, Reagents for 50 tests, color chart and accessories in a box, 0 • 1 • 3 • 5 • 10 • 15 • 20 mg/L CTAB, Bromophenol blue, Visual, 239×135×47 mm (Including packaging), 50, 1
Katalog # MANA931051
Leverandør:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Detergents
Katalog # MANA931051
Leverandør:  MACHEREY-NAGEL
CAS nummer:  
VISOCOLOR® Colorimetric Test, Detergents
VISOCOLOR® Colorimetric Test, Detergents

VISOCOLOR® ECO Cationic detergents, Reagents for 50 tests, color chart and accessories in a box, 0 • 1 • 3 • 5 • 10 • 15 • 20 mg/L CTAB, Bromophenol blue, Visual, 239×135×47 mm (Including packaging), 50, 1

CAS nummer:  
Price
1.300,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Reagents for the determination of detergents in water samples.

  • Combines simple implementation with high measurement reliability

Simple determination directly on site or in the laboratory.

Sea water analysis: Yes

Hazardous material: Yes

Specifikationer

Specifikationer

  • Kemisk metode:
    Bromophenol blue
  • Pakn:
    1
  • Beskrivelse:
    VISOCOLOR® ECO Cationic detergents
  • Dimensioner:
    239×135×47 mm (Including packaging)
  • Til enhed:
    Visual
  • Test pr. sæt:
    50
  • Område:
    0 • 1 • 3 • 5 • 10 • 15 • 20 mg/L CTAB
  • Indhold:
    Reagents for 50 tests, color chart and accessories in a box
  • Cat. No.:
    MANA931051
  • Platform:
    Colorimetric test kits
  • Holdbarhed (måneder):
    24
  • Vægt:
    510 g (Including packaging)
  • Model:
    MANA931051
  • Opbevaringstemperatur:
    15...25 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.