Snap ring caps for snap ring vials, N 11
PE, hard, transparent, center hole, 45° shore A, Red rubber/FEP colourless, 1,0 mm
Katalog # MANA702731
Leverandør: MACHEREY-NAGEL
Gå til:
Product Details & Documents
Ready assembled snap ring closures made of polyethylene are available for the snap ring vials.
- Available as hard cap version in transparent and further colours or as soft cap in light blue for easy handling
- Large choice of septa materials
- Most frequent version are closures with cross-slit silicone/PTFE septum, which supports easy penetration of blunt HPLC needles
Documents
MACHEREY-NAGEL Vials and Caps.pdf
Yderligere produktdetaljer
Specifikationer
- Hårdhed:45° shore A
- Septum:Red rubber/FEP colourless
- Model:PE, hard, transparent, center hole
- Seq:01
- Tykkelse:1,0 mm