Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Snap ring caps for snap ring vials, N 11

Snap ring caps for snap ring vials, N 11

PE, hard, blue, center hole, 50° shore A, PTFE red/silicone white/PTFE red, 1,0 mm
Katalog # MANA702718.1
Leverandør:   MACHEREY-NAGEL
Snap ring caps for snap ring vials, N 11
Katalog # MANA702718.1
Leverandør:  MACHEREY-NAGEL
Snap ring caps for snap ring vials, N 11
Snap ring caps for snap ring vials, N 11

PE, hard, blue, center hole, 50° shore A, PTFE red/silicone white/PTFE red, 1,0 mm

Price
282,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Ready assembled snap ring closures made of polyethylene are available for the snap ring vials.

  • Available as hard cap version in transparent and further colours or as soft cap in light blue for easy handling
  • Large choice of septa materials
  • Most frequent version are closures with cross-slit silicone/PTFE septum, which supports easy penetration of blunt HPLC needles

Documents

Specifikationer
  • Hårdhed:
    50° shore A
  • Septum:
    PTFE red/silicone white/PTFE red
  • Model:
    PE, hard, blue, center hole
  • Seq:
    12
  • Tykkelse:
    1,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.