Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Snap ring caps for snap ring vials, N 11

Snap ring caps for snap ring vials, N 11

PE, soft, light blue, center hole, 45° shore A, Red rubber/FEP colourless, 1,0 mm
Katalog # MANA702063.2080
Leverandør:   MACHEREY-NAGEL
Snap ring caps for snap ring vials, N 11
Katalog # MANA702063.2080
Leverandør:  MACHEREY-NAGEL
Snap ring caps for snap ring vials, N 11
Snap ring caps for snap ring vials, N 11

PE, soft, light blue, center hole, 45° shore A, Red rubber/FEP colourless, 1,0 mm

Price
180,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents

Ready assembled snap ring closures made of polyethylene are available for the snap ring vials.

  • Available as hard cap version in transparent and further colours or as soft cap in light blue for easy handling
  • Large choice of septa materials
  • Most frequent version are closures with cross-slit silicone/PTFE septum, which supports easy penetration of blunt HPLC needles

Documents

Specifikationer
  • Hårdhed:
    45° shore A
  • Septum:
    Red rubber/FEP colourless
  • Model:
    PE, soft, light blue, center hole
  • Seq:
    13
  • Tykkelse:
    1,0 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.